MilliporeSigma
  • Increased loss of brain DNA in the neonatal vasopressin-deficient Brattleboro rat, but not in normal rat treated with vasopressin antagonist.
Increased loss of brain DNA in the neonatal vasopressin-deficient Brattleboro rat, but not in normal rat treated with vasopressin antagonist.
Neuroscience letters (1993-06-25)
G J Boer, F G Snijdewint, R Licht, P te Riele
ABSTRACT
In order to establish whether vasopressin (VP) influences brain cell survival, [3H]thymidine was injected in 10-day-old vasopressin-deficient Brattleboro rat pups, as well as in Wistar pups treated, neonatally, with the VP antagonist dP[Tyr(Me)2]VP followed by subsequent measurement of [3H]DNA in olfactory bulbs and cerebellum days and weeks thereafter. Results show, first of all, that the incorporation of [3H]thymidine into DNA was enhanced in the homozygous (HOM) Brattleboro, when compared with the heterozygous (HET; non-vasopressin-deficient) controls. The difference is due to the greater and prolonged tissue availability of [3H]thymidine, possibly pointing to an altered thymidine uptake and/or metabolism. Between postnatal days 25 and 39 no differences were seen in [3H]DNA content of the brain parts of the HET and Wistar control rats. For the HOM rats, however, a loss of [3H]DNA was seen (up to 8%), indicating that increased postnatal brain cell death might occur in the mutant. The antagonist treatment in Wistar rat up to 21 days of age failed to show a similar effect. It is proposed that general growth impairments, rather than VP receptor-mediated effects, lead to the brain cell loss.