MilliporeSigma
  • [Optical isomer analysis of ethylamphetamine from human urine and an anorectic drug, Apetinil Depo].
[Optical isomer analysis of ethylamphetamine from human urine and an anorectic drug, Apetinil Depo].
Nihon hoigaku zasshi = The Japanese journal of legal medicine (1993-08-01)
T Nagai, S Kamiyama, K Matsushima
ABSTRACT
Two analyzed cases of Ethylamphetamine (EAMP) were reported. By using HPLC with a chiral column, racemic (d, l)-EAMP were detected from the urine of a Japanese male and an anorectic drug, which had been possessed by a Thai club hostess. From the urine of the former, d-MAMP, d-AMP and l-MAP were also detected. EAMP is now categorized to be one of psychotropic drugs in Japan, but not under the strict control of Psychostimulants Law. In near future, because of the similar action to stimulant drugs, EAMP will be apprehensive of the abuse in Japan.