MilliporeSigma
  • Inhibition of 3,5,3'-triiodothyronine binding to its receptor in rat liver by protease inhibitors and substrates.
Inhibition of 3,5,3'-triiodothyronine binding to its receptor in rat liver by protease inhibitors and substrates.
Molecular and cellular endocrinology (1993-05-01)
J Brtko, J Knopp, M E Baker
ABSTRACT
Various protease inhibitors (e.g. phenylmethanesulfonyl fluoride (PMSF), tosyl-phenylalanine chloromethyl ketone (TosPheCH2Cl)) and substrates (e.g., tosyl-arginine methyl ester (TosArgOMe), tryptophan methyl ester (TrpOMe)) inhibit the binding of adrenal and sex steroids to their cognate receptors (Hubbard and Kalimi (1985) Mol. Cell. Biochem. 66, 101-109). Here we extend this finding to the receptor for 3,5,3'-triiodothyronine (T3) in rat liver nuclei. We find that PMSF, TosPheCH2Cl and other protease inhibitors as well as TosArgOMe, TrpOMe, tyrosine methyl ester (TyrOMe) and tyrosine ethyl ester (TyrOEt) inhibit binding of 125I-T3 to its receptor in rat liver nuclei. Inhibition by protease substrates appears to be at or close to the hormone binding domain. By analogy with the known mechanism of binding of protease inhibitors and substrates to enzymes, we suggest that the T3 receptor contains a nucleophilic site at or close to the hormone binding domain.
Sigma-Aldrich
L-Tyrosine methyl ester, 98%
Sigma-Aldrich
L-Tyrosine methyl ester hydrochloride