MilliporeSigma
  • Continuous electrophoretic separations in narrow channels coupled to small-bore capillaries.
Continuous electrophoretic separations in narrow channels coupled to small-bore capillaries.
Analytical chemistry (1993-11-15)
J M Mesaros, G Luo, J Roeraade, A G Ewing
ABSTRACT
Continuous zone electrophoretic separations in channels have been demonstrated. This new technique has the potential to continuously sample and separate analytes from volume-limited microenvironments. A small-bore capillary is used to electrophoretically sample, but not separate, a mixture of dansylated amino acids. The capillary is coupled to a quartz channel structure in a manner which allows continuous injection of the sampled material into the channel. The channel functions to continuously separate the sampled material via electrophoresis. A laser-induced fluorescence detection scheme, which involves two fiber optic arrays situated at the channel exit, monitors eluting analytes. A continuous separation of dansylated amino acids on the time scale of a few minutes demonstrates the utility of the technique. Sampling has been performed continuously up to 400 s, and initial detection limits are approximately 30 microM.