MilliporeSigma
  • Skin contamination, airborne concentrations, and urinary metabolite excretion of propoxur during harvesting of flowers in greenhouses.
Skin contamination, airborne concentrations, and urinary metabolite excretion of propoxur during harvesting of flowers in greenhouses.
American journal of industrial medicine (1993-11-01)
R Brouwer, K van Maarleveld, L Ravensberg, W Meuling, W de Kort, J J van Hemmen
ABSTRACT
In eight greenhouses used for carnation culture, workers engaged in harvesting (n = 16), were monitored for dermal and respiratory exposure and urinary excretion of propoxur. Dermal exposure of hands and forearms was estimated from dislodgable foliar residue, using a transfer factor (a measure of transfer of pesticides from leaves to the skin) and the total number of working hours. Total estimated dermal and respiratory exposure during harvesting ranged from 0.2 to 46 mg and from 3 to 278 micrograms, respectively. To study the relationship between external and internal exposure to propoxur, respiratory and dermal exposure levels were compared with the total amount of 2-isopropoxyphenol (IPP), the major metabolite of propoxur, excreted in urine in 24 hr. The Pearson correlation coefficient between dermal exposure and the total amount of excreted IPP was 0.95. A correlation coefficient of 0.84 was found between respiratory exposure and the amount of IPP excreted. The latter association was probably caused by the covariation of respiratory and dermal exposure levels (r = 0.85). Assuming negligible oral absorption, calculations indicated that dermal exposure could account for > 80% of the amount of excreted IPP. On the basis of the amount of IPP excreted, there was no reason to suspect increased health risks for workers from exposure to propoxur during harvesting.