MilliporeSigma
Lipoprotein patterns in acrylamide gel electrophoresis.
Science (New York, N.Y.) (1966-01-21)
S Raymond, J L Miles, J C Lee
ABSTRACT
Lipoproteins of serum stained before electrophoresis gave distinct patterns in acrylamide gel of 3-percent concentration. Samples compared in parallel on gel slabs showed qualitative differences in migration rates of beta-lipoprotein bands.