MilliporeSigma
Genetic toxicology testing of di(2-ethylhexyl) terephthalate.
Environmental and molecular mutagenesis (1994-01-01)
E D Barber
ABSTRACT
Di(2-ethylhexyl) terephthalate (DEHT) is a commercially produced chemical (Kodaflex DOTP) that is used as a general purpose, low-volatility plasticizer for polyvinyl chloride and other polymeric materials. Less than 30 million kilograms of DEHT are produced annually. DEHT is isomeric with di(2-ethylhexyl) phthalate (DEHP), a nongenotoxic rodent carcinogen whose mode of action has been suggested to derive from its ability to produce hepatocellular proliferation and/or hepatic peroxisome proliferation. Thus it is important to know the behavior of DEHT in genotoxicity assays in order to compare it with that of DEHP and other phthalate ester plasticizers. It is known from previously published studies that rats fed DEHT in the diet at 2,000 mg/kg produce urine that is negative in the Ames Salmonella bacterial mutagenicity assay in the presence and absence of induced rat liver S-9 and in the presence and absence of beta-glucuronidase/aryl sulfatase. Reported here are the results of direct testing of DEHT in the Ames plate incorporation assay, the Chinese hamster ovary/hypoxanthine guanine phosphoribosyl transferase (CHO/HGPRT) in vitro mammalian cell mutagenicity assay, and an in vitro chromosome aberrations assay using CHO cells. The results for mono(ethylhexyl) terephthalate (MEHT), a metabolite of DEHT, in the Ames Salmonella bacterial mutagenicity assay are also presented. All test results for both DEHT and MEHT were found to be negative, and it is therefore concluded that DEHT, like its isomeric relative DEHP, is not genotoxic.
Sigma-Aldrich
Dioctyl terephthalate, ≥96%