MilliporeSigma
Selective opioid dipeptides.
Biochemical and biophysical research communications (1994-02-15)
P A Temussi, S Salvadori, P Amodeo, C Bianchi, R Guerrini, R Tomatis, L H Lazarus, D Picone, T Tancredi
ABSTRACT
The surprising change of selectivity induced by the change of chirality in peptides containing the tetrahydro-3-isoquinoline carboxylic acid (Tic) in second position, interpreted as a conformational preference induced on the Tyr-Xaa-Phe domain, can instead be attributed to the Tyr-Tic message domain. The relative spatial disposition of the aromatic ring of delta-selective non peptidic opiates is compatible with a message domain, in opioid peptides, of only two residues. This hypothesis was tested through the synthesis of Tyr-L-Tic-NH2, Tyr-D-Tic-NH2, Tyr-L-Tic-Ala-NH2, Tyr-L-Tic-Ala-OH and Tyr-D-Tic-Ala-NH2. Peptides containing Tyr-L-Tic- behave as very selective delta antagonists and those containing Tyr-DTic- as non selective agonists. This is the first case of opioid peptides containing a two-residue message domain and of opioid dipeptides with substantial opioid activity.
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%