MilliporeSigma
  • Modulation by isoproterenol and propranolol of somatostatin receptors in synaptosomes from rat frontoparietal cortex.
Modulation by isoproterenol and propranolol of somatostatin receptors in synaptosomes from rat frontoparietal cortex.
Brain research (1993-06-18)
S López-Sañudo, E Arilla
ABSTRACT
DL-Propranolol (PRO), a beta-adrenergic blocking agent, and the neuropeptide somatostatin (SS) have central nervous system depressant and anticonvulsive properties. To investigate a possible relationship between these two components, we studied the influence of PRO and DL-isoproterenol (ISO), a beta-adrenergic agonist, on the somatostatinergic system in the rat frontoparietal cortex. The short- (5 h) and long-term (14 days) administration of ISO (5 mg/kg, intraperitoneally (i.p.)), or of PRO (10 mg/kg, i.p.) did not affect somatostatin-like immunoreactivity (SLI) content in the frontoparietal cortex of male Wistar rats. Both short- and long-term ISO administration decreased the number of specific [125I]Tyr11-SS receptors in synaptosomes from frontoparietal cortex (31%, P < 0.05, and 26%, P < 0.02, after short- and long-term administration, respectively) without changing the affinity constant. This decrease in the number of [125I]Tyr11-SS receptors was not due to a direct effect of ISO on these receptors since no decrease in binding was produced by high concentrations of ISO (10(-5) M) when added in vitro. This decrease could be blocked by pretreatment with PRO. Short- and long-term administration of PRO alone produced an increase in the [125I]Tyr11-SS binding in frontoparietal cortex (26%, P < 0.02, and 40%, P < 0.001, after short- or long-term administration, respectively) without changing the affinity constant.