MilliporeSigma
  • Metabolic studies on haloperidol and its tetrahydropyridine analog in C57BL/6 mice.
Metabolic studies on haloperidol and its tetrahydropyridine analog in C57BL/6 mice.
Chemical research in toxicology (1994-05-01)
C J Van der Schyf, K Castagnoli, E Usuki, H G Fouda, J M Rimoldi, N Castagnoli
ABSTRACT
The neuroleptic agent haloperidol (HP) is biotransformed in humans to a pyridinium metabolite, HPP+, that displays neurotoxic properties resembling those of the 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-derived neurotoxic pyridinium metabolite MPP+. We report here that HP and its tetrahydropyridine dehydration product 4-(4-chlorophenyl)-1-[4-(4-fluorophenyl)-4-oxobutyl]-1,2,3,6- tetrahydropyridine (HPTP) are metabolized in vivo by the MPTP-susceptible C57BL/6 mouse to several pyridinium metabolites including HPP+ and the 4-(4-chlorophenyl)-1-[4-(4-fluorophenyl)-4-hydroxybutyl]pyridinium species RHPP+, the pyridinium species corresponding to reduced haloperidol (RHP), a major circulating metabolite of HP. Atmospheric pressure ion-spray (API) mass spectral data also suggest the formation of fluorophenyl ring-hydroxylated derivatives of these two pyridinium metabolites. Furthermore, HPLC tracings reveal the presence of HPP+, RHPP+, and two phenolic pyridinium metabolites in brain tissue extracts of HPTP, but not HP, treated mice. The neurotoxic potential of MPTP-type pyridinium species suggests that these metabolites may contribute to some of the neurological disorders observed in humans undergoing chronic HP treatment.
Product Description
Supelco
Haloperidol metabolite II, analytical standard