MilliporeSigma
  • Influence of chromium picolinate on glucose usage and metabolic criteria in growing Holstein calves.
Influence of chromium picolinate on glucose usage and metabolic criteria in growing Holstein calves.
Journal of animal science (1994-06-01)
L D Bunting, J M Fernandez, D L Thompson, L L Southern
ABSTRACT
Holstein calves, 10 steers (98 +/- 16 kg; Exp. 1) and 14 heifers (122 +/- 7 kg; Exp. 2), were fed either corn-cottonseed hull basal diets (CON; 15.1 and 13.2% CP, respectively) or CON plus 370 micrograms/kg of Cr as Cr-tripicolinate (CrP). After either 58 (Exp. 1) or 56 (Exp. 2) days of ADG and DMI measurement, calves were individually fed 1.4% of their BW every 12 h through the end of each experiment. Nitrogen balance, i.v. glucose tolerance tests (IVGTT), i.v. insulin challenge tests (IVICT), baseline growth hormone (GH) measurements, and GH-releasing hormone challenges were then performed on calves in split groups on d 70 through 87 in Exp. 1. In Exp. 2, all challenges began on d 78 and N balance was not determined. Inclusion of CrP did not affect (P > .10) ADG, DMI, or ADG/DMI of pooled data from Exp. 1 and 2 or N balance criteria in Exp. 1. Plasma glucose, insulin, and NEFA concentrations were not affected (P > .10) by CrP. Total plasma cholesterol was lower (P < .05) for CrP- than for CON-fed calves at wk 4 in Exp. 1 and wk 6 in Exp. 2. During IVGTT, calves fed CrP vs CON had higher (P < .05) clearance rates for glucose in Exp. 1 (2.68 vs 1.88%/min) and in Exp. 2 (2.64 vs 2.08%/min). During IVICT, calves fed CrP vs CON had higher clearance rates for glucose in Exp. 1 (2.98 vs 2.34%/min; P = .12) and in Exp. 2 (2.89 vs 2.00%/min; P < .05).(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Chromium picolinate, ≥98% (HPLC)