MilliporeSigma
  • Myocardial salvage by chimyl alcohol: possible role of peroxisomal dysfunction in reperfusion injury.
Myocardial salvage by chimyl alcohol: possible role of peroxisomal dysfunction in reperfusion injury.
Annals of the New York Academy of Sciences (1994-06-17)
N Maulik, A Tosaki, R M Engelman, G A Cordis, D K Das
ABSTRACT
The results of this study suggest that reperfusion of ischemic myocardium may lead to the peroxisomal disorder both functionally and biochemically. An alkyl glycerol such as chimyl alcohol can protect the ischemic heart from the reperfusion injury probably by enhancing the plasmalogen synthesis.