MilliporeSigma
  • Determination of codeine and its metabolites in microsomal incubates by high-performance liquid chromatography.
Determination of codeine and its metabolites in microsomal incubates by high-performance liquid chromatography.
Journal of chromatography. B, Biomedical applications (1994-02-18)
M Pawula, P N Shaw, D A Barrett
ABSTRACT
A rapid and sensitive HPLC method has been developed for the determination of codeine, norcodeine and morphine in small volumes of a biological matrix, using a cyanopropyl column and a combination of coulometric and UV detection. The compounds were isolated using C18 solid-phase extraction cartridges prior to quantitative analysis. The limit of detection was 250 pg/ml for morphine and 5 ng/ml for both norcodeine and codeine. Recovery of each compound was greater than 90% and intra- and inter-assay precision was better than 10%. The method has been used to study the metabolism of codeine in microsomal incubations.
Product Description
Supelco
Norcodeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.