MilliporeSigma
  • Coincidental appearance of the alpha 1 subunit of the GABA-A receptor and the type I benzodiazepine receptor near birth in macaque monkey visual cortex.
Coincidental appearance of the alpha 1 subunit of the GABA-A receptor and the type I benzodiazepine receptor near birth in macaque monkey visual cortex.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience (1994-06-01)
A Hendrickson, D March, G Richards, A Erickson, C Shaw
ABSTRACT
The expression of subtypes of the GABA-A/benzodiazepine receptor complex has been studied during pre- and postnatal development of Macaca monkey visual cortex using complementary radioligand and immunocytochemical labeling. Type I benzodiazepine receptors were labeled directly by [3H]zolpidem. Type II receptors were determined by the amount of binding for [3H]flunitrazepam (FZ) persisting in the presence of the type I-specific ligand CL218872. Monoclonal antibody bd24 was used to label alpha 1 subunits and bd17 to label beta 2 and beta 3 subunits of the GABA-A receptor. Radioligand binding data and bd17 immunoreactivity indicated that type II benzodiazepine receptors were present by fetal day (Fd) 74 (44% of gestation). Immunoreactivity for the beta 2/beta 3 subunits increased until 3-6 weeks after birth, and then declined somewhat into adulthood. Neither radioligand labeling for type I receptors nor immunocytochemical staining for the alpha 1 subunit were apparent until mid-gestation. Both markers appeared shortly before birth in layer 4C, and then in other cortical layers after birth. Immunoreactivity for the alpha 1 subunit increased steadily after birth until it became more intense than that for beta 2/3 subunits in the adult. Quantitative densitometry of CL218872 competition for [3H]FZ binding showed that type I/II distribution was 22%/78% at Fd103; 42%/58% at Fd131; 67%/33% at 9 months; and 61%/39% in adult visual cortex. This "switch" between benzodiazepine receptor subtypes overlaps the postnatal critical period for geniculostriate development, suggesting that the change from type II to type I receptors and the appearance of alpha 1 subunits may play a decisive role in the maturation of geniculocortical axon terminations and cortical response properties. It remains to be shown whether this "switch" is dependent on functional visual input.