MilliporeSigma
  • A pharmacological profile of the novel, peripherally-selective kappa-opioid receptor agonist, EMD 61753.
A pharmacological profile of the novel, peripherally-selective kappa-opioid receptor agonist, EMD 61753.
British journal of pharmacology (1994-12-01)
A Barber, G D Bartoszyk, H M Bender, R Gottschlich, H E Greiner, J Harting, F Mauler, K O Minck, R D Murray, M Simon
ABSTRACT
1. The pharmacological properties of the novel diarylacetamide kappa-opioid receptor agonist, EMD 61753, have been compared with those of ICI 197067 (a centrally-acting kappa agonist) and ICI 204448 (a peripherally-selective kappa agonist). 2. EMD 61753 binds with high affinity (IC50 5.6 nM) and selectivity (kappa:mu:delta:sigma binding ratio 1:536:125: > 1,786) to kappa-opioid receptors and is a full and potent (IC50 54.5 nM) agonist in an in vitro assay for kappa-opioid receptors (rabbit vas deferens preparation). 3. Systemically-applied [14C]-EMD 61753 is found in high concentrations in the lungs, liver, adrenal glands and kidneys. Considerably less radioactivity is detected in the whole brain, and this radioactivity is concentrated in the region of the cerebral ventricles in the choroid plexuses. EMD 61753 penetrates only poorly into the CNS. 4. EMD 61753 was weakly effective in pharmacological tests of central activity. This compound reversed haloperidolol-induced DOPA accumulation in the nucleus accumbens of the rat only at a dose of 30 mg kg-1, s.c., (doses of 0.1, 1.0 and 10 mg kg-1, s.c., and 1.0, 10 and 100 mg kg-1, p.o., were inactive). Hexobarbitone-induced sleeping in mice was prolonged by EMD 61753 at threshold doses of 10 mg kg-1, s.c., and 100 mg kg-1, p.o., whereas the motor performance of rats in the rotarod test was impaired by EMD 61753 with an ID50 value of 453 mg kg-1, s.c. 5. EMD 61753 produced dose-dependent, naloxone-reversible antinociception in the mouse formalin test (1st phase ID50 1.9 mg kg-1, s.c., and 10.4 mg kg-1, p.o.; 2nd phase ID50 0.26 mg kg-1, s.c., and 3.5 mg kg-1, p.o.) and rodent abdominal constriction test (ID50 mouse 1.75 mg kg-1, s.c., and 8.4 mg kg-1, p.o.; ID50 rat 3.2 mg kg-1, s.c., and 250 mg kg-1, p.o.). EMD 61753 was inactive, or only weakly effective, in the rat pressure test under normalgesic conditions. After the induction of hyperalgesia with carrageenin, however, this compound elicited potent, dose-dependent (ID50 0.08 mg kg-1, s.c., and 6.9 mg kg-1, p.o., after remedial application, and 0.2 mg kg-1, s.c., and 3.1 mg kg-1, p.o., after prophylactic application) and naloxone-reversible antinociception. The antinociceptive action of systemically-applied (50 mg kg-1, p.o.) EMD 61753 in the hyperalgesic pressure test was completely inhibited by injection of the K-opioid antagonist norbinaltorphimine (100 Lg) into the inflamed tissue, a result which indicates that this opioid effect is mediated peripherally.6. Cutaneous plasma protein extravasation produced by antidromic electrical stimulation of the rat saphenous nerve was dose-dependently inhibited by systemically-applied EMD 61753 (ID13 values 3.7 mg kg-1, s.c., and 35.8 mg kg-1, p.o.), and this effect was completely antagonized by intraplantar application of norbinaltorphimine (50 microg). Extravasation elicited by the intraplantar application of substance P (10 microg) was not influenced by the administration of EMD 61753.7. EMD 61753 produced dose-dependent diuresis in non-hydrated rats at doses of and above 1.0 mg kg-1, s.c., and 10 mg kg-1, p.o., and in saline-loaded rats at doses of and above 10 mg kg-1, s.c.,and 30mgkg-1, p.o.8. The prostaglandin-mediated fall in mean arterial blood pressure elicited in anaesthetized rats by i.v.application of arachidonic acid was not inhibited by prior treatment with EMD 61753 (10mg kg-1,p.o.). Thus, a blockade of prostaglandin synthesis via inhibition of cyclo-oxygenase activity does not contribute to the in vivo effects of EMD 61753 and its metabolites.9 The present experiments therefore indicate that EMD 61753 is a potent, selective and orally-effective full ic-opioid receptor agonist which has a limited ability to penetrate the blood-brain barrier and elicit centrally-mediated sedation, putative aversion, diuresis, and antinociception. The inhibitory actions of systemically-applied EMD 61753 against hyperalgesic pressure nociception and neurogenic inflammation are mediated peripherally, probably by opioid receptors on the endings of sensory nerve fibres.