MilliporeSigma
  • [A clinical evaluation of SY5555 in the treatment of pediatric infections].
[A clinical evaluation of SY5555 in the treatment of pediatric infections].
The Japanese journal of antibiotics (1995-02-01)
S Furukawa, T Okada
ABSTRACT
1. SY5555 dry syrup (powder which is dissolved before use) was administered to 25 patients with bacterial infections (6 cases of bronchitis, 2 cases of bronchopneumonia, 1 case of pertussis, 3 cases of scarlet fever, 5 cases of tonsillitis, 3 cases of urinary tract infections, 2 cases of staphylococcal scalded skin syndrome, 1 case of impetigo, 2 cases of purulent lymphadenitis). 2. Clinical efficacies were excellent in 11 patients and good in 13, poor in 1 with an efficacy rate of 96.0%. As pathogenic organisms, 15 strains were identified and 14 of them were eradicated with eradication rate of 93.3%. 3. No side effects were observed. As for abnormal laboratory test results increase in eosinophiles in 2 cases, decrease in filamented neutrophiles in 1 case, elevation of GOT and GTP in 1 case and elevation of GPT and gamma-GTP were observed. 4. There was no rejection incidence of the drug during the therapy. From the above results, we consider SY5555 in dry syrup form to be a useful and safe drug in the treatment of various bacterial infections in pediatric patients.
Sigma-Aldrich
Faropenem sodium hydrate, ≥98% (HPLC)