MilliporeSigma
  • NMDA receptor subtype selectivity: eliprodil, polyamine spider toxins, dextromethorphan, and desipramine selectively block NMDA-evoked striatal acetylcholine but not spermidine release.
NMDA receptor subtype selectivity: eliprodil, polyamine spider toxins, dextromethorphan, and desipramine selectively block NMDA-evoked striatal acetylcholine but not spermidine release.
Journal of neurochemistry (1995-05-01)
M Nankai, D Fage, C Carter
ABSTRACT
NMDA receptor stimulation concomitantly increases the release of [14C]acetylcholine and [3H]-spermidine from rat striatal slices in vitro. The NMDA-induced release of both acetylcholine and spermidine was blocked with equal potency by the NMDA channel blocker phencyclidine (0.1-10 microM). However, certain other channel blockers, including dextromethorphan (1-100 microM), which antagonized NMDA-evoked acetylcholine release without affecting NMDA-evoked spermidine release, and dextrorphan (1-100 microM) and memantine (1-100 microM), which block NMDA-evoked acetylcholine release more potently than NMDA-evoked spermidine release, showed greater selectivity of action. As previously shown for ifenprodil, eliprodil (SL82.0715; 1-100 microM) blocked NMDA-evoked acetylcholine but not spermidine release. This selectivity is also observed for other agents interacting with the polyamine site(s) on the NMDA receptor, including arcaine (1-1,000 microM), philanthotoxin343, and argiotoxin636 (10 microM) and was also noted for desipramine (1-100 microM). The NMDA-induced release of acetylcholine and spermidine is likely to be mediated by different native NMDA receptor subtypes, and several NMDA antagonists may be candidates for a selective action at a particular NMDA receptor subtype.