MilliporeSigma
  • Highly specific oxidative cross-linking of proteins mediated by a nickel-peptide complex.
Highly specific oxidative cross-linking of proteins mediated by a nickel-peptide complex.
Biochemistry (1995-04-11)
K C Brown, S H Yang, T Kodadek
ABSTRACT
The Ni(II) complex of the tripeptide NH2-Gly-Gly-His-COOH is shown to mediate efficient protein-protein cross-linking in the presence of oxidants such as oxone and monoperoxyphthalic acid. Only proteins that associate specifically in solution are cross-linked under these conditions. Preliminary probes of the mechanism of the reaction suggest that the active intermediate may be a high-valent metal complex that attacks aromatic amino acids.
Sigma-Aldrich
Gly-Gly-His