MilliporeSigma
  • Comparison of the sensitivity of Salmonella typhimurium strains YG1024 and YG1012 for detecting the mutagenicity of aromatic amines and nitroarenes.
Comparison of the sensitivity of Salmonella typhimurium strains YG1024 and YG1012 for detecting the mutagenicity of aromatic amines and nitroarenes.
Mutation research (1993-01-01)
M Watanabe, T Sofuni, T Nohmi
ABSTRACT
Salmonella typhimurium YG1024 is a derivative of S. typhimurium TA98 with a high level of N-hydroxyarylamine O-acetyltransferase (OAT) activity. We have demonstrated that this strain is highly sensitive to the mutagenic actions of N-hydroxyarylamines derived from aromatic amines and nitroarenes. In this paper, we compared the sensitivities of YG1024 with those of S. typhimurium YG1012, which has about 4 times higher OAT activity than YG1024 but lacks plasmid pKM101. It turned out that YG1024 was more sensitive to the mutagenic actions of 1-aminonaphthalene, 1-nitropyrene, 1,8-dinitropyrene and 2-nitronaphthalene than YG1012 and showed comparable sensitivity to 2-hydroxy-acetylaminofluorene, 2-aminoanthracene and 2-amino-6-methyldipyrido[1,2-a:3',2'-d]imidazole (Glu-P-1) to YG1012. These results suggested that YG1024 is more suitable than YG1012 for the efficient detection of mutagenic aromatic amines and nitroarenes.