MilliporeSigma
  • Quantitative cytochemistry of nuclear and cytoplasmic proteins using the Naphthol Yellow S and dinitrofluorobenzene staining methods.
Quantitative cytochemistry of nuclear and cytoplasmic proteins using the Naphthol Yellow S and dinitrofluorobenzene staining methods.
The Histochemical journal (1981-09-01)
J Tas, J James
ABSTRACT
The 'total protein staining' of biological specimens with the electrostatically binding Naphthol Yellow S or the covalently binding dinitrofluorobenzene must be interpreted as methods which yield data on the specific amino acid pool of the proteins concerned. Both dyes bind to certain free amino-acid side-chains, giving different dye--protein ratios for various proteins. In the presence of DNA, dinitrofluorobenzene stains all proteins present in cell nuclei, whereas Naphthol Yellow S only stains the majority of the non-histone proteins. When protein staining methods are combined with the Feulgen--Pararosanile (SO2) procedure for DNA, decreased Feulgen--DNA contents were measured in dinitrofluorobenzene-stained isolated nuclei and lymphocytes.
Product Description
Supelco
Naphthol Yellow S, analytical standard
Sigma-Aldrich
Naphthol Yellow S, for microscopy (Hist.), for the precipitation (of amino acids and peptides)