MilliporeSigma
  • Synthesis of alpha and beta-glycopyranosyl-serine derivatives by enzymic transglycosylation.
Synthesis of alpha and beta-glycopyranosyl-serine derivatives by enzymic transglycosylation.
Biomedica biochimica acta (1991-01-01)
D Cantacuzene, S Attal, S Bay
ABSTRACT
The transglycosylation from raffinose and lactose to Aloc-Ser-OMe is catalyzed respectively by alpha and beta galactosidases. Transglycosylation from cellobiose has been achieved with beta-glucosidase. The simplicity of the enzymatic synthesis, the stereospecificity of the condensations in one-pot reactions and the ease of purification give the method value for large scale preparation of beta-linked derivatives. The protective groups of the serine residue can be cleaved under mild conditions: the ester group has been removed quantitatively by papain catalyzed hydrolysis and the Aloc group by a Pd (0) hydrostannolytic cleavage.
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized, powder, ≥4 U/mg