MilliporeSigma
  • Comparison of liquid and agar-solidified defined media regarding the physiological mechanism by which beta-2-thienylalanine inhibits growth of Escherichia, Shigella, and Salmonella cultures.
Comparison of liquid and agar-solidified defined media regarding the physiological mechanism by which beta-2-thienylalanine inhibits growth of Escherichia, Shigella, and Salmonella cultures.
Microbiology and immunology (1980-01-01)
K J Brown, G W Tannock, R B Elliott, D R Lines
ABSTRACT
Growth comparisons were made, using Shigella, Escherichia, and Salmonella cultures, in liquid and agar-solidified defined media containing beta-2-thienylalanine (beta-2-t). The comparisons were performed to determine the nature of growth inhibition by beta-2-t under different physical growth conditions. In a plate assay, with increasing beta-2-t mixed into the agar, inhibition of Escherichia and Shigella increased. However, Salmonella cultures were not inhibited even at the highest beta-2-t concentrations used. With beta-2-t added to liquid cultures, however, dose-response growth relationships were exhibited by all three genera. The differences occurring in beta-2-t inhibition between liquid and plate assay conditions were not due to composition of culture plates, time of challenge of cultures with beta-2-t, availability of oxygen and associated differences in ratios of volume of media to available surface area, selection of mutants in the plate assay, or to extractable substances from the agar. However, when beta-2-t diffusion into the liquid medium was delayed by using agar plug diffusion cultures, a physiological mechanism was demonstrable which largely protected Salmonella cultures, but not Escherichia and Shigella cultures, from growth inhibition.
Sigma-Aldrich
3-(2-Thienyl)-L-alanine, ≥98.0% (TLC)
Sigma-Aldrich
3-(2-Thienyl)-DL-alanine, ≥98%