MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Seasonal variations in HIOMT activity during the night in the pineal gland of 21 day old male Wistar rats.
Seasonal variations in HIOMT activity during the night in the pineal gland of 21 day old male Wistar rats.
Journal of neural transmission (1980-01-01)
M G Balemans, F A Bary, W C Legerstee, J Van Benthem
ABSTRACT
In the pineal of 21-day old male Wistar rats hydroxyindole-O-methyltransferase (HIOMT) activities involved in the synthesis of several 5-methoxyindoles were determined during the night in April, June, October and January. A high HIOMT activity for the synthesis of melatonin/5-methoxytryptophol was determined in the months of January and April. In June and October a decrease was observed. The activity maxima coincide with peaks of activity found for the synthesis of 5-methoxytryptophan. Synthesis of 5-methoxytryptamine occurred only in June and October, whereas the synthesis of 5-methoxyindole-3-acetic acid occurred only in January. From these results it may be concluded that January and April are the most active months of those tested for the melatonin/5-methoxytryptophol synthesis in the rat pineal gland. A possible physiological role of the 5-methoxyindoles other than melatonin and 5-methoxytryptophol is discussed.