MilliporeSigma
  • The enzymatic status of circulating lymphocytes as an index of the regenerative process in the lungs under stimulation with pyrimidine and purine derivatives. A clinical experimental study.
The enzymatic status of circulating lymphocytes as an index of the regenerative process in the lungs under stimulation with pyrimidine and purine derivatives. A clinical experimental study.
Haematologia (1982-01-01)
G L Bilich, L V Nazarova, E V Sungurova
ABSTRACT
Enzyme activities in the circulating lymphocytes permit to judge the character of the regenerative process in the lung, and the severity of the patient's condition; it helps in estimating the prognosis and in evaluating the efficacy of the applied therapy. Potassium orotate and Riboxin intensify the regenerative process in the lung undergoing a compensatory-hypertrophic rearrangement, and enhance the functional activity of circulating lymphocytes as participants of the regenerative process and stimulate the oxidation-reduction processes in lymphocytes.
Sigma-Aldrich
1-Methyluracil, 99%