MilliporeSigma
  • Jack bean urease (EC 3.5.1.5). V. On the mechanism of action of urease on urea, formamide, acetamide, N-methylurea, and related compounds.
Jack bean urease (EC 3.5.1.5). V. On the mechanism of action of urease on urea, formamide, acetamide, N-methylurea, and related compounds.
Canadian journal of biochemistry (1980-12-01)
N E Dixon, P W Riddles, C Gazzola, R L Blakeley, B Zerner
ABSTRACT
Acetamide and N-methylurea have been shown for the first time to be substrates for jack bean urease. In the enzymatic hydrolysis of urea, formamide, acetamide, and N-methylurea at pH 7.0 and 38 degrees C, kcat has the values 5870, 85, 0.55, and 0.075 s-1, respectively. The urease-catalyzed hydrolysis of all these substrates involves the active-site nickel ion(s). Enzymatic hydrolysis of the following compounds could not be detected: phenyl formate, p-nitroformanilide, trifluoroacetamide, p-nitrophenyl carbamate, thiourea, and O-methylisouronium ion. In the enzymatic hydrolysis of urea, the pH dependence of kcat between pH 3.4 and 7.8 indicates that at least two prototropic forms are active. Enzymatic hydrolysis of urea in the presence of methanol gave no detectable methyl carbamate. A mechanism of action for urease is proposed which involves initially an O-bonded complex between urea and an active-site Ni2+ ion and subsequently an O-bonded carbamato-enzyme intermediate.
Sigma-Aldrich
N-(4-Nitrophenyl)formamide, 97%
Sigma-Aldrich
o-Methylisourea bisulfate, 99%
Sigma-Aldrich
O-Methylisourea hemisulfate salt, 99%