MilliporeSigma
  • Trimipramine kinetics and absolute bioavailability: use of gas-liquid chromatography with nitrogen-phosphorus detection.
Trimipramine kinetics and absolute bioavailability: use of gas-liquid chromatography with nitrogen-phosphorus detection.
Clinical pharmacology and therapeutics (1984-03-01)
D R Abernethy, D J Greenblatt, R I Shader
ABSTRACT
Kinetic parameters were derived from trimipramine and desmethyltrimipramine plasma concentrations after administration of intravenous (12.5 mg) and oral (50 mg) trimipramine in nine subjects. Elimination t1/2 after intravenous dosing was (mean +/- SE) 23 +/- 1.9 hr. Volume of distribution by the area method was 30.9 +/- 3.5 l/kg and total metabolic clearance was 15.9 +/- 1.5 ml/min/kg. Plasma protein binding of trimipramine, as determined by equilibrium dialysis, averaged 94.9%, with a range of 93.8% to 96.4%. Peak plasma level attained was 28.2 +/- 4.4 ng/ml at 3.1 +/- 0.6 hr after oral dosing. Absolute bioavailability was 41.4% +/- 4.4% (range of 17.8% to 62.7%). These data indicate that trimipramine has incomplete and variable systemic availability, that it is more highly protein bound than other tricyclic antidepressants, and, on the basis of its elimination t1/2, that it could be administered on a twice-daily basis without marked interdose fluctuations in plasma levels.
Product Description
Supelco
N-Desmethyltrimipramine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®