MilliporeSigma
Tissue response to anodyne medicaments.
Oral surgery, oral medicine, and oral pathology (1984-11-01)
F Barnett, C Duran, G Hasselgren, L Tronstad
ABSTRACT
The tissue-irritating effect of anodyne medicaments was studied via enzyme histochemical evaluation of muscle tissue (screening test) and pulp tissue (usage test). Of the medicaments tested, formocresol was the most irritating, followed by Cresatin, eugenol, and oil of pimenta leaf. The effects of Cresatin, eugenol, and oil of pimenta leaf were not statistically distinguishable.
Sigma-Aldrich
m-Tolyl acetate, 97%