MilliporeSigma
Three new contaminants detected in postmortem blood samples.
Journal of analytical toxicology (1984-11-01)
J Vine, T R Watson, B Ford
ABSTRACT
Three contaminants were identified during drug screening of postmortem blood samples which had been stored in glass bottles with a black rubber seal. Two of these contaminants, cyanoethyl dimethyldithiocarbamate and N-phenyl-2-naphthylamine, were found to come from the rubber seal of some bottles. The third contaminant was not a single compound but rather a mixture of aryl phosphates with a composition very similar to technical grade tritolyl phosphate. The origin of these phosphates is at present unknown.
Sigma-Aldrich
N-Phenyl-2-naphthylamine, 97%