MilliporeSigma
  • Characterization of a mitochondrial transport system for branched chain alpha-keto acids.
Characterization of a mitochondrial transport system for branched chain alpha-keto acids.
The Journal of biological chemistry (1985-11-15)
S M Hutson, S L Rannels
ABSTRACT
Efflux of branched chain alpha-keto acids from preloaded rat heart mitochondria was slow at low external pH. Efflux was first order, and measured rate constants, kappa efflux, were 0.104 +/- 0.005 and 0.115 +/- 0.006 min-1 for alpha-ketoisovalerate and alpha-ketoisocaproate (KIC), respectively. Efflux was stimulated significantly by branched chain alpha-keto acids and related carboxylates such as alpha-ketocaproate and alpha-ketovalerate, but not by substrates for the pyruvate transporter. KIC was the preferred substrate, and the apparent exchange K0.5 for KIC was 0.14 +/- 0.10 mM. Exchange was 7-8-fold faster than efflux, and the maximal rate of exchange at saturating concentrations of alpha-ketoisovalerate and KIC appeared to be independent of the metabolite used. It is proposed that branched chain alpha-keto acids cross the inner mitochondrial membrane on a specific transporter. Transport occurs with a proton, i.e. by proton symport, and is sensitive to inhibition by cinnamic acid derivatives.