MilliporeSigma
  • A study of serum antibody activity in workers with occupational exposure to diphenylmethane diisocyanate.
A study of serum antibody activity in workers with occupational exposure to diphenylmethane diisocyanate.
Allergy (1985-07-01)
K S Tse, A Johnson, H Chan, M Chan-Yeung
ABSTRACT
The prevalence of sensitization was studied in a group of 76 foundry workers with occupational exposure to diphenylmethane diisocyanate (MDI). Ten workers had clinical evidence of asthma, 40 had non-asthmatic respiratory symptoms, and 26 were asymptomatic. Specific IgE antibodies to MDI were found in two workers (2.6%) and specific IgG antibodies, in five workers (6.6%). The prevalence of IgE and IgG antibodies was higher in the 10 subjects with asthma than in the non-asthmatic group. The prevalence of anti-paratolyl-monoisocyanate antibodies was not significantly different from that of anti-MDI antibodies, and both haptenic determinants displayed a high degree of cross-reactivity in the RAST inhibition test. The role of humoral immunological mechanisms in MDI-induced asthma is unclear in view of the rather low prevalence of these serum antibodies in this group of workers.
Sigma-Aldrich
p-Tolyl isocyanate, 99%