MilliporeSigma
  • Synthesis of [18F]2-deoxy-2-fluoro-D-glucose from highly reactive [18F]tetraethylammonium fluoride prepared by hydrolysis of [18F]fluorotrimethylsilane.
Synthesis of [18F]2-deoxy-2-fluoro-D-glucose from highly reactive [18F]tetraethylammonium fluoride prepared by hydrolysis of [18F]fluorotrimethylsilane.
The International journal of applied radiation and isotopes (1985-05-01)
L G Hutchins, A L Bosch, M S Rosenthal, R J Nickles, S J Gatley
ABSTRACT
18F-Labeled fluorotrimethylsilane was prepared by nucleophilic substitution of chlorotrimethylsilane with reactor produced [18F]fluoride. Hydrolysis of fluorotrimethylsilane by aqueous tetraethylammonium hydroxide followed by removal of water with a mechanical pump gave a powerful source of no carrier added nucleophilic 18F. Reaction of this purified 18F preparation with 4,6-benzylidene-1-beta-O-methyl D-mannopyranoside-2.3-cyclic sulfate was complete in 2 min at 80 degrees C and gave two labeled products with similar retention times on reverse phase HPLC. Allowing for decay and handling losses during deprotection, the maximum yield of [18F]2-deoxy-2-fluoro-D-glucose from no-carrier added tetraethylammonium fluoride was 50%. Incorporation of 18F into organic products was 30% complete in 10 min at room temperature. An identical time-course was observed for reaction of 3-O-triflyl-1,2-5,6-diisopropylidene-D-allofuranose, the starting material for 3-deoxy-3-fluoro-D-glucose. Reaction of tetraethylammonium fluoride with chlorotrimethylsilane was more rapid and much more tolerant of water than the fluorosugar reactions. Chlorotrimethylsilane can be used to recover unreacted 18F from reaction mixtures.