MilliporeSigma
  • Determination of 23 polycyclic aromatic hydrocarbons in atmospheric particulate matter of the Paris area and photolysis by sunlight.
Determination of 23 polycyclic aromatic hydrocarbons in atmospheric particulate matter of the Paris area and photolysis by sunlight.
International journal of environmental analytical chemistry (1985-01-01)
B Muel, S Saguem
ABSTRACT
Twenty-three polycyclic aromatic hydrocarbons (PAH) were determined in atmospheric particulate matter in 4 places of the Paris area at several times of the year. Fractionation was performed by reversed-phase high-pressure liquid chromatography. Determination was done by recording emission or excitation fluorescence spectra via a stopped-flow technique. Triphenylene was also extemporaneously determined by its phosphorescence spectrum at low temperature. Among the PAH determined dibenz(e,ghi)perylene has not been detected before in atmospheric particulate matter. The 10 more abundant PAH ranged from 0.1 to 40 ng/m3 of filtered air. Concentrations in August are from 14 to 250 times less than in January depending on the PAH. The reasons for this difference of behaviour among the PAH were investigated with regard to their photochemical and non-photochemical reactivity.
Product Description
Supelco
Indeno[1,2,3-cd]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Indeno[1,2,3-cd]pyrene, analytical standard
Dibenzo[a,e]pyrene, BCR®, certified reference material