MilliporeSigma
  • Chromogenic labeling of monosaccharides using 4'-N,N-dimethylamino-4-aminoazobenzene.
Chromogenic labeling of monosaccharides using 4'-N,N-dimethylamino-4-aminoazobenzene.
Analytical biochemistry (1985-05-15)
G Rosenfelder, M Mörgelin, J Y Chang, C A Schönenberger, D G Braun, H Towbin
ABSTRACT
Twenty-three monosaccharides, e.g., D- or L-pentoses, D- or L-hexoses, heptose, 2- or 6-deoxyhexoses, 2-deoxy-2-aminohexoses, hexuronic acids, and N-acetylmuramic acid, were coupled to the azo dye 4'-N,N-dimethylamino-4-aminoazobenzene by reductive amination using sodium cyanoborohydride as reducing agent and in the presence of pentaerythritol. The structure of the colored glycamines was established by mass spectrometry. The average yield of the reaction was more than 80%. The sugar derivatives were separated either by silica-gel thin-layer chromatography or by high-performance liquid chromatography. Spectrophotometric quantitation was performed in the visible range at the picomole level. The method was applied to the determination of the sugar composition of the glycosphingolipid globotetraosyl ceramide and the human milk oligosaccharide lacto-N-fucopentaose I.
Sigma-Aldrich
N,N-Dimethyl-4,4′-azodianiline, 97%