MilliporeSigma
  • Liquid chromatographic determination of noxytiolin and 1-methyl-2-thiourea in serum: application to pharmacokinetic studies in rabbits and humans.
Liquid chromatographic determination of noxytiolin and 1-methyl-2-thiourea in serum: application to pharmacokinetic studies in rabbits and humans.
Journal of pharmaceutical sciences (1985-02-01)
D Debruyne, M A Moulin, H Bricard, M C Bigot
ABSTRACT
A high-performance liquid chromatographic method is described which determines noxytiolin and 1-methyl-2-thiourea concentrations in serum. Valid determination requires immediate ultracentrifugation of blood samples, rapid serum freezing, and injection into the chromatograph within 6 h. A number of pharmacokinetic parameters were calculated from serum concentration data in rabbits and humans. An unknown metabolite was detected in both species but its structure was not identified.
Sigma-Aldrich
N-Methylthiourea, 97%