MilliporeSigma
Dipeptide analysis in human urine.
Journal of chromatography (1986-10-31)
J Jandke, G Spiteller
ABSTRACT
Fractions of dipeptides, obtained from human urine by a combination of cation-exchange chromatography, ligand-exchange chromatography and reversed-phase chromatography, were transformed into their N-heptafluorobutyryl methyl ester derivatives and then subjected to capillary gas chromatography. The profiles obtained indicate the presence of many dipeptides in human urine. For the first time, alpha-Asp-Hyp, Pro-Phe and gamma-Glu-Phe were detected in the urine of healthy individuals.
Sigma-Aldrich
Prolylphenylalanine, ≥95% (HPLC)