MilliporeSigma
  • Cantharidin biosynthesis in a blister beetle: inhibition by 6-fluoromevalonate causes chemical disarmament.
Cantharidin biosynthesis in a blister beetle: inhibition by 6-fluoromevalonate causes chemical disarmament.
Experientia (1986-07-15)
J E Carrel, J P Doom, J P McCormick
ABSTRACT
Biosynthesis of cantharidin in a blister beetle, Lytta polita, is effectively inhibited by 6-fluoromevalonate. Inhibition is attributed specifically to the fluorine substituent. Biochemical inhibition has not been demonstrated previously for an arthropod's defensive substance.
Sigma-Aldrich
6-Fluoromevalonate, ≥90% (GC), viscous liquid