MilliporeSigma
  • Carcinogen-DNA adduct formation in the lungs and livers of preweanling CD-1 male mice following administration of [3H]-6-nitrochrysene, [3H]-6-aminochrysene, and [3H]-1,6-dinitropyrene.
Carcinogen-DNA adduct formation in the lungs and livers of preweanling CD-1 male mice following administration of [3H]-6-nitrochrysene, [3H]-6-aminochrysene, and [3H]-1,6-dinitropyrene.
Cancer research (1987-12-01)
K B Delclos, R P Walker, K L Dooley, P P Fu, F F Kadlubar
ABSTRACT
6-Nitrochrysene (NC) and 6-aminochrysene (AC) have been shown to be potent lung and liver carcinogens when administered in multiple i.p. doses to preweanling mice. 1,6-Dinitropyrene has been shown to be a strong hepatocarcinogen but a weak lung carcinogen in this same bioassay. We have examined carcinogen-DNA adduct profiles in the target tissues of preweanling male CD-1 mice following administration of single or multiple doses of these compounds. Depending on the tissue and the dosing schedule, the total level of DNA modification in animals dosed with [3H]NC was 2- to 9-fold higher than in animals dosed with [3H]AC. Regardless of the dosing schedule, DNA isolated from the lungs and livers of both [3H]NC- and [3H]AC-treated preweanling male mice contained a single major and chromatographically identical adduct. This major adduct, which accounted for as much as 90% of the total carcinogen-DNA adducts in enzymatic hydrolysates from treated animals, was chromatographically distinct from the major C8-purine-substituted adducts formed from the reaction of N-hydroxy-AC with calf thymus DNA. In contrast to the results obtained with NC and AC, the major carcinogen-DNA adduct formed in the livers of mice treated with [3H]-1,6-dinitropyrene was found to cochromatograph with 1-N-(deoxyguanosin-8-yl)amino-6-nitropyrene, a product derived from N-hydroxy-1-amino-6-nitropyrene. Since NC and its nitro-reduced derivative, AC, yielded an identical carcinogen-DNA adduct in vivo and this adduct was not derived from N-hydroxy-AC, we conclude that the metabolic activation of NC in the neonatal mouse must involve some previously undescribed combination of ring-oxidation and nitro-reduction pathways. This activation pathway could be an important factor in determining the potency of NC and AC as carcinogens in this bioassay system.
Sigma-Aldrich
6-Aminochrysene, 95%