MilliporeSigma
  • Horse serum butyrylcholinesterase kinetics: a molecular mechanism based on inhibition studies with dansylaminoethyltrimethylammonium.
Horse serum butyrylcholinesterase kinetics: a molecular mechanism based on inhibition studies with dansylaminoethyltrimethylammonium.
Biochemistry and cell biology = Biochimie et biologie cellulaire (1987-06-01)
G Cauet, A Friboulet, D Thomas
ABSTRACT
The kinetics of the hydrolysis of butyrylthiocholine by horse serum butyrylcholinesterase (acylcholine acylhydrolase; BuChE; EC 3.1.1.8) exhibit an activation phenomenon at high substrate concentrations. At least two mechanistic models can account for the enzyme kinetics: one assumes the binding of an additional substrate molecule on the acyl-enzyme intermediate, and the other hypothesizes the existence of a peripheral regulatory site for the substrate. (1-Dimethylaminonaphthalene-5-sulfonamidoethyl)-trimethylammonium perchlorate, a potent reversible inhibitor, appears to affect BuChE activity by binding to a peripheral site. The inhibition is of the mixed type at low substrate concentrations and of the competitive type at high substrate concentrations. This is consistent with a peripheral site for the binding of the substrate responsible for the activation phenomenon.