MilliporeSigma
  • Acetylation, deacetylation and hydroxylation of sulphamethoxazole and N4-acetylsulphamethoxazole in the turtle Pseudemys scripta elegans.
Acetylation, deacetylation and hydroxylation of sulphamethoxazole and N4-acetylsulphamethoxazole in the turtle Pseudemys scripta elegans.
The Veterinary quarterly (1987-10-01)
T B Vree, J B Vree, Y A Hekster, J F Nouws
ABSTRACT
The turtle Pseudemys scripta elegans is able to hydroxylate and acetylate Sulphamethoxazole in a way that is comparable to man, i.e. the rate and yield of hydroxylation equals that of the acetylation. The hydroxy metabolites 5-hydroxy- and N4-acetyl-5-hydroxysulphamethoxazole are not glucuronidated. N4-acetylsulphamethoxazole is neither deacetylated nor hydroxylated.
Product Description
Sulfamethoxazole impurity A, European Pharmacopoeia (EP) Reference Standard