MilliporeSigma
  • Gas chromatographic determination of residue levels of the herbicides trifluralin, benefin, ethalfluralin, and isopropalin in soil with confirmation by mass selective detection.
Gas chromatographic determination of residue levels of the herbicides trifluralin, benefin, ethalfluralin, and isopropalin in soil with confirmation by mass selective detection.
Journal - Association of Official Analytical Chemists (1988-11-01)
S D West, J H Weston, E W Day
ABSTRACT
A method is presented for the simultaneous or individual determination of the dinitroaniline herbicides trifluralin, benefin, ethalfluralin, and isopropalin in soil. The herbicides are extracted with acetonitrile-water (99 + 1), and the extracts are purified with small, disposable Florisil cartridges prior to analysis by gas chromatography using an electron capture detector or a mass selective detector. When electron capture detection is used as the primary detection system, confirmation with selective detection can be obtained using gas chromatography-mass spectrometry with a mass selective detector and a capillary column operated in the split mode. The limit of detection is 0.01 ppm, and recoveries averaged 95-112% for the 4 herbicides in several different soil types fortified at levels of 0.01-0.33 ppm.
Product Description
Supelco
Ethalfluralin, PESTANAL®, analytical standard
Supelco
Benfluralin, PESTANAL®, analytical standard