MilliporeSigma
  • Neonatal treatment with vasopressin antagonist dP[Tyr(Me)2]AVP, but not with vasopressin antagonist d(CH2)5[Tyr(Me)2]AVP, inhibits body and brain development and induces polyuria in the rat.
Neonatal treatment with vasopressin antagonist dP[Tyr(Me)2]AVP, but not with vasopressin antagonist d(CH2)5[Tyr(Me)2]AVP, inhibits body and brain development and induces polyuria in the rat.
Neurotoxicology and teratology (1988-07-01)
F G Snijdewint, G J Boer
ABSTRACT
Two vasopressin antagonists, d(CH2)5[Tyr(Me)2]AVP and dP[Tyr(Me)2]AVP, were given to Wistar rats from postnatal day 1 to 21 in order to investigate the influence on development and later diuresis. The latter antagonist significantly reduced body growth from day 3 postnatally onwards. At postnatal day 35 body, total brain, cerebellar and kidney weights were significantly reduced compared with controls. Diuresis, measured at one month of age, was four- to five-fold higher than the control group. Combined treatment with vasopressin failed to abolish the weight disturbances or polyuria. However, animals treated with the vasopressin antagonist d(CH2)5[Tyr(Me)2]AVP did not show developmental or diuretic deficits. Allometric analysis of brain/body relationship of the young animals indicated a disturbance of brain development by dP[Tyr(Me)2]AVP. Although the body and brain growth retardation induced by dP[Tyr(Me)2]AVP supports the hypothesis of a role for vasopressin in brain ontogeny, it can also be the result of a nonAVP-related toxic effect, since it could not be prevented by concomitant treatment with vasopressin.