MilliporeSigma
  • Tumor-selective metabolism of 5-fluoro-2'-deoxycytidine coadministered with tetrahydrouridine compared to 5-fluorouracil in mice bearing Lewis lung carcinoma.
Tumor-selective metabolism of 5-fluoro-2'-deoxycytidine coadministered with tetrahydrouridine compared to 5-fluorouracil in mice bearing Lewis lung carcinoma.
Cancer research (1987-05-01)
D A Boothman, T V Briggle, S Greer
ABSTRACT
The metabolic products formed and incorporated into the nucleic acids (RNA and DNA) of mice bearing Lewis lung carcinoma (LLC) following optimal doses of 5-fluorouracil (FUra), 5-fluoro-2'-deoxyuridine (FdUrd), and 5-fluoro-2'-deoxycytidine (FdCyd) coadministered with tetrahydrouridine (H4Urd), a potent inhibitor of cytidine deaminase, were examined. Treatment with FdCyd plus H4Urd resulted in a tumor-selective incorporation and formation of antimetabolites compared to either FUra or FdUrd treatments. Between 45- and greater than 5400-fold higher levels of the potent thymidylate synthetase inhibitor, 5-fluoro-2'-deoxyuridylate (FdUMP), were formed in tumor than in any of the normal tissues analyzed. RNA-level antimetabolites (FUra, 5-fluorouridine, and 5-fluorouridylate) were also between 3 and greater than 990-fold higher in tumor compared to normal tissue following FdCyd plus H4Urd administration. DNA-level antimetabolites (FdCyd, 5-fluorodeoxycytidylate, FdUrd, and FdUMP) were from 2- to 6-fold higher in tumor compared to normal tissue. FUra and FdUrd treatments resulted in between 3 and greater than 1300-fold higher RNA-level antimetabolites and from 4 to greater than 1020-fold higher FdUMP pools in normal tissues than FdCyd plus H4Urd treatment. DNA-level antimetabolites were also from 4- to 32-fold higher in normal tissues following optimal doses of FUra or FdUrd. In tumor tissue, optimal doses of FUra or FdUrd resulted in lower (a) FdUMP levels (5- to 2-fold), (b) RNA-level antimetabolites (6- to 3-fold), and (c) DNA-level antimetabolites (10- to 4-fold) compared to an optimal dosage of FdCyd plus H4Urd. In serum, the administration of H4Urd resulted in the protection of FdCyd from systemic catabolism, unlike that found with FUra or FdUrd. Substantial levels of FdUMP, FUrd, and FUMP were noted in serum following FUra or FdUrd treatment. The formation of di- and triphosphate antimetabolite pools and the incorporation of antimetabolites into the RNA and DNA of normal and tumor tissues demonstrated trends similar to those mentioned above with nucleoside, mononucleotide, and free base pools. H4Urd treatment of 25 mg/kg did not affect the elevated levels of deoxycytidine kinase or deoxycytidylate deaminase in LLC tumor tissue or the low levels found in normal tissue. A critical feature of this chemotherapeutic strategy using FdCyd plus H4Urd was that the elevated level of cytidine deaminase in LLC tumor tissue was inhibited less than 10% by the administration of 25 mg/kg H4Urd, whereas deoxycytidine deaminase activities in normal tissues (including bone marrow and intestine) were inhibited greater than 93%.(ABSTRACT TRUNCATED AT 400 WORDS)
Sigma-Aldrich
5−Fluoro−2′−deoxycytidine, ≥98% (HPLC), powder