MilliporeSigma
  • Synthesis of decadeoxyribonucleotides containing 5-modified uracils and their interactions with restriction endonucleases Bgl II, Sau 3AI and Mbo I (nucleosides and nucleotides 82).
Synthesis of decadeoxyribonucleotides containing 5-modified uracils and their interactions with restriction endonucleases Bgl II, Sau 3AI and Mbo I (nucleosides and nucleotides 82).
Nucleic acids research (1988-06-10)
T Hayakawa, A Ono, T Ueda
ABSTRACT
Decadeoxyribonucleotides containing uracil, 5-bromouracil, 5-cyanouracil and 5-ethyluracil in recognition sequences of restriction endonucleases Bgl II, Sau 3AI, Mbo I were synthesized. Decanucleotides containing 5-bromouracil in place of thymine had essentially the same susceptibility to all the restriction endonucleases. Uracil-containing decanucleotides were however very resistant to attack. Decanucleotides containing 5-cyanouracil in the recognition sequence were strongly resistant to hydrolysis by Sau 3AI, but were hydrolysed by Bgl II and Mbo I as well as the parent decanucleotide. Decanucleotides containing 5-ethyluracil were strongly resistant to hydrolysis by Sau 3AI, but were partially resistant to hydrolysis by Bgl II and Mbo I.