MilliporeSigma
  • Rapid determination of internal volumes of membrane vesicles with electron spin resonance-stopped flow technique.
Rapid determination of internal volumes of membrane vesicles with electron spin resonance-stopped flow technique.
Biochimica et biophysica acta (1988-01-13)
K Anzai, K Higashi, Y Kirino
ABSTRACT
We have developed an electron spin resonance (ESR)-stopped flow technique and employed it for the simple and rapid determination of internal volumes of biomembrane vesicles and liposomes. A vesicle suspension containing a neutral and membrane-permeable spin label, 2,2,6,6-tetramethyl-4-oxopiperidine-1-oxyl (TEMPONE), was mixed in the stopped-flow apparatus with an isotonic solution of relatively impermeable line broadening agents, potassium tris(oxalato)chromate(III) or potassium ferricyanide, and an ESR spectrum was recorded. From the relative intensity of the sharp triplet signal due to TEMPONE in the aqueous space within vesicles, the determination of the internal aqueous volume was straightforward. Using this technique, it is possible to measure intravesicular volumes in 0.1 s. The internal volume of sonicated phospholipid vesicles was approximately 0.3 microliter/mg lipid. The light fraction of sarcoplasmic reticulum membrane vesicles isolated from rabbit skeletal muscle was estimated to have an internal volume of 2.2-2.6 microliter/mg protein in its resting state. Activation of Ca2+ pumps in the membrane upon addition of ATP and Ca2+ ions decreased the internal volume by about 10%. This finding supports the hypothesis that the Ca2+ pump is electrogenic and that the efflux of potassium ions compensates for the influx of positive charges. The present technique is widely applicable to the simple and rapid determination of the internal volumes of membrane vesicles.
Sigma-Aldrich
Potassium chromium(III) oxalate trihydrate, 98%