MilliporeSigma
  • Effects of the lipolysis inhibiting agent beta-pyridylcarbinol (Ronicol) in acute myocardial ischaemia.
Effects of the lipolysis inhibiting agent beta-pyridylcarbinol (Ronicol) in acute myocardial ischaemia.
Acta physiologica Hungarica (1989-01-01)
T Fazekas, L Szekeres
ABSTRACT
The antilipolytic, nicotinic acid analogue beta-pyridylcarbinol (Ronicol) has previously been reported to decrease the free fatty acid (FFA) concentration of the arteria-blood, and to moderate the FFA-uptake and O2-consumption of the myocardium; on this basis, the drug may be expected to exert a cardioprotective action. The cardiac effects of Ronicol were therefore studied on a self-control, 'single-vessel' coronary artery ligature dog model. The left anterior descending coronary artery (LAD) was prepared in the in situ heart of anaesthetized, thoracotomized animals. Following the control ligation, a stabilization period and Ronicol infusion (1 mg/kg iv. during 10 minutes), the LAD was repeatedly ligated. The duration of the individual occlusions was 10 minutes. Ronicol significantly decreased the arterial FFA concentration and the epicardial ST segment elevation; its antilipolytic and anti-ischaemic effects were protracted and were still observed 120 minutes after pretreatment. The drug did not decrease the inhomogeneity of ventricular depolarization in the ischaemic myocardium and in the dose applied it had no influence on the heart rate, arterial blood pressure, left ventricular end-diastolic pressure and left ventricular contractility (LV dP/dtmax). In the canine myocardial infarction model employed it was observed that the duration of the anti-ischaemic effect of Ronicol (1 mg/kg iv.) is about 120 minutes. It has the advantage that it does not possess the unwanted cardiovascular side-effects displayed by nicotinic acid observed by us too in this model earlier (Cardiol. Hung. 13, 33-41, 1984).
Sigma-Aldrich
3-Pyridinemethanol, 98%