MilliporeSigma
Evidence for age-related change in plasma 19-hydroxyandrostenedione.
Endocrinologia japonica (1989-12-01)
K Higuchi, A Ogo, T Maki, M Haji, R Takayanagi, M Ohashi, H Nawata, K Kato, H Ibayashi
ABSTRACT
The steroid, 19-hydroxyandrost-4-ene-3, 17-dione (19-hydroxyandrostene-dione, 19-OH-A-dione) has been known to enhance the mineralocorticoid action of aldosterone. To investigate the age-related change in the plasma 19-OH-A-dione concentration, plasma 19-OH-A-dione, androst-4-ene-3, 17-dione (A-dione), aldosterone and cortisol of 38 non-hypertensive healthy subjects (18 young men and 20 aged men) measured by specific radioimmunoassays. The basal plasma 19-OH-A-dione and A-dione concentration in aged men was significantly lower than in young men (P less than 0.01). Moreover, there was found to be a positive correlation between plasma 19-OH-A-dione and A-dione (P less than 0.01). On the other hand, plasma aldosterone and cortisol in aged men showed a tendency to decrease, but no statistical significance compared to young men was observed. This study demonstrated that there was an apparent age-related decrease not only in plasma A-dione, but also in plasma 19-OH-A-dione, an amplifier or aldosterone action.