MilliporeSigma
  • Glutamate receptor subtypes may be classified into two major categories: a study on Xenopus oocytes injected with rat brain mRNA.
Glutamate receptor subtypes may be classified into two major categories: a study on Xenopus oocytes injected with rat brain mRNA.
Neuron (1989-07-01)
H Sugiyama, I Ito, M Watanabe
ABSTRACT
Three major subtypes of glutamate receptors that are coupled to cation channels are known. Recently an additional subtype that is coupled to G proteins and stimulates inositol phospholipid metabolism (the metabotropic glutamate receptor) has been proposed. The pharmacological characteristics of this receptor have now been examined. Although it shares some agonists with N-methyl-D-aspartate- and quisqualate-subtype receptors, it shares virtually no antagonists with any of the three cation channel-coupled receptor subtypes. Thus the metabotropic glutamate receptor belongs to a receptor category that is completely different from that of the other three receptor subtypes, not only functionally, but also pharmacologically.