MilliporeSigma
  • Suloctidil increases the rat brain cortex microvascular regeneration after a lesion.
Suloctidil increases the rat brain cortex microvascular regeneration after a lesion.
Life sciences (1989-01-01)
F De Paermentier, P Heuschling, B Knoops, P Janssens De Varebeke, G Pauwels, C Laszlo De Kaszon-Jakabfalva, P Van den Bosch De Aguilar
ABSTRACT
A "cavity" lesion made by aspiration in the rat occipital cortex induces a parenchymal and a vascular reaction in its vicinity. The first was mainly characterized by cellular necrosis and gliosis, the second by an increase of the vascular network. In vehicle treated rats, a 50% significant increase of the vascular network was observed around the cavity 4 days after the lesion, in comparison to the uninjured contralateral cortex. The effects of a vasoactive substance, suloctidil, on the vascular reaction was studied in the brain cortex. A single oral dose of suloctidil (30 mg/kg; 2 hours before the sacrifice) gave the same effect as the vehicle group. After 8 days of suloctidil oral administration (30 mg/kg; twice daily: 4 days before lesion and 4 days after) a significant increase (123%) of the vascular network was observed around the cavity. The hypothetical ways by which a chronic treatment of suloctidil induces this increase of the neovascularization observed after cortical lesion are discussed.