MilliporeSigma
  • Concurrent naltrexone and prolonged exposure therapy for patients with comorbid alcohol dependence and PTSD: a randomized clinical trial.
Concurrent naltrexone and prolonged exposure therapy for patients with comorbid alcohol dependence and PTSD: a randomized clinical trial.
JAMA (2013-08-09)
Edna B Foa, David A Yusko, Carmen P McLean, Michael K Suvak, Donald A Bux, David Oslin, Charles P O'Brien, Patricia Imms, David S Riggs, Joseph Volpicelli
ABSTRACT
Alcohol dependence comorbid with posttraumatic stress disorder (PTSD) has been found to be resistant to treatment. In addition, there is a concern that prolonged exposure therapy for PTSD may exacerbate alcohol use. To compare the efficacy of an evidence-based treatment for alcohol dependence (naltrexone) plus an evidence-based treatment for PTSD (prolonged exposure therapy), their combination, and supportive counseling. A single-blind, randomized clinical trial of 165 participants with PTSD and alcohol dependence conducted at the University of Pennsylvania and the Philadelphia Veterans Administration. Participant enrollment began on February 8, 2001, and ended on June 25, 2009. Data collection was completed on August 12, 2010. Participants were randomly assigned to (1) prolonged exposure therapy plus naltrexone (100 mg/d), (2) prolonged exposure therapy plus pill placebo, (3) supportive counseling plus naltrexone (100 mg/d), or (4) supportive counseling plus pill placebo. Prolonged exposure therapy was composed of 12 weekly 90-minute sessions followed by 6 biweekly sessions. All participants received supportive counseling. The Timeline Follow-Back Interview and the PTSD Symptom Severity Interview were used to assess the percentage of days drinking alcohol and PTSD severity, respectively, and the Penn Alcohol Craving Scale was used to assess alcohol craving. Independent evaluations occurred prior to treatment (week 0), at posttreatment (week 24), and at 6 months after treatment discontinuation (week 52). Participants in all 4 treatment groups had large reductions in the percentage of days drinking (mean change, -63.9% [95% CI, -73.6% to -54.2%] for prolonged exposure therapy plus naltrexone; -63.9% [95% CI, -73.9% to -53.8%] for prolonged exposure therapy plus placebo; -69.9% [95% CI, -78.7% to -61.2%] for supportive counseling plus naltrexone; and -61.0% [95% CI, -68.9% to -53.0%] for supportive counseling plus placebo). However, those who received naltrexone had lower percentages of days drinking than those who received placebo (mean difference, 7.93%; P = .008). There was also a reduction in PTSD symptoms in all 4 groups, but the main effect of prolonged exposure therapy was not statistically significant. Six months after the end of treatment, participants in all 4 groups had increases in percentage of days drinking. However, those in the prolonged exposure therapy plus naltrexone group had the smallest increases. In this study of patients with alcohol dependence and PTSD, naltrexone treatment resulted in a decrease in the percentage of days drinking. Prolonged exposure therapy was not associated with an exacerbation of alcohol use disorder. clinicaltrials.gov Identifier: NCT00006489.
Product Description
Supelco
Naltrexone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®