MilliporeSigma
  • Simultaneous determination of dextropropoxyphene, norpropoxyphene and methaqualone in plasma by gas chromatography with selective nitrogen detection.
Simultaneous determination of dextropropoxyphene, norpropoxyphene and methaqualone in plasma by gas chromatography with selective nitrogen detection.
Journal de toxicologie clinique et experimentale (1990-03-01)
P Kintz, A Tracqui, P Mangin, A A Lugnier, A A Chaumont
ABSTRACT
A method for the identification and quantification of dextropropoxyphene, norpropoxyphene, and methaqualone in plasma by GC/NPD is presented. The procedure employs cyclizine as the internal standard and requires no derivatization. After a single-step extraction, analysis is achieved in 8 min. The lower limits of detection were found to be 6, 12, and 1 ng/ml for dextropropoxyphene, norpropoxyphene, and methaqualone, respectively. This method appears to be rapid, sensitive, and applicable to forensic and clinical toxicological analyses.